Zirconium standard solution, for ICP, in 5% HCl, CRM, VHG
Lieferant: Promochem
PROOVHG-PZRH-100EA 45 EUR
PROOVHG-PZRH-100 PROOVHG-PZRH-500 PROOVHG-TZRH-100 PROOVHG-PZRH-50 PROOVHG-TZRH-500
Zirconium standard solution, for ICP, in 5% HCl, CRM, VHG
Standards Kationischer Standards Zirkonium-Standards
These solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Standards: Traceable to NIST, SRM.