Tungsten standard solution, for ICP, in H₂O, CRM, VHG
Lieferant: Promochem
PROOVHG-PWW-500EA 121 EUR
PROOVHG-PWW-500 PROOVHG-PWW-100 PROOVHG-TWW-100 PROOVHG-TWW-500 PROOVHG-PWW-50
Tungsten standard solution, for ICP, in H₂O, CRM, VHG
Standards Kationischer Standards Wolfram-Standards
These solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Standards: Traceable to NIST, SRM.