Vanadium standard solution, for ICP, in HNO₃, CRM, VHG
Lieferant: LGC Standards
PROOVHG-TVN-100EA 89 EUR
PROOVHG-TVN-100 PROOVHG-PVN-500 PROOVHG-PVN-50 PROOVHG-LVN-100 PROOVHG-PVN-100 PROOVHG-TVN-500
Vanadium standard solution, for ICP, in HNO₃, CRM, VHG
Standards Kationischer Standards Vanadium-Standards
These solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Standards: Traceable to NIST, SRM.