Samarium standard solution, for ICP, in 5% HNO₃, CRM, VHG
Lieferant: LGC Standards
VHG
PROOVHG-TSMN-500EA 270 EUR
PROOVHG-TSMN-500 PROOVHG-PSMN-500 PROOVHG-PSMN-100 PROOVHG-TSMN-100
Samarium standard solution, for ICP, in 5% HNO₃, CRM, VHG
Standards Kationischer Standards Samarium-Standards
These solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Standards: Traceable to NIST, SRM.