Mercury standard solution, for ICP, in HNO₃, CRM, VHG
Lieferant: LGC Standards
PROOVHG-THGN-500EA 193 EUR
PROOVHG-THGN-500 PROOVHG-LHGN-100 PROOVHG-THGN-100 PROOVHG-PHGN-100 PROOVHG-LHGN-250 PROOVHG-PHGN-500
Mercury standard solution, for ICP, in HNO₃, CRM, VHG
Standards Kationischer Standards Quecksilber-Standards
These solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Standards: Traceable to NIST, SRM.