Iron standard solution, for ICP, in 5% HNO₃, VHG
Lieferant: Promochem
PROOVHG-TFEN-100EA 79 EUR
PROOVHG-TFEN-100 PROOVHG-PFEN-50 PROOVHG-TFEN-500 PROOVHG-PFEN-500 PROOVHG-PFEN-100
Iron standard solution, for ICP, in 5% HNO₃, VHG
Standards Kationischer Standards Eisen-Standards
The solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Standards: Traceable to NIST, SRM.