Chromium standard solution, for ICP, in 5% HCl, VHG
Lieferant: Promochem
PROOVHG-TCRH-100EA 83 EUR
PROOVHG-TCRH-100 PROOVHG-PCRH-500 PROOVHG-TCRH-500 PROOVHG-PCRH-100
Chromium standard solution, for ICP, in 5% HCl, VHG
Standards Kationischer Standards Chrom-Standards
The solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Standards: Traceable to NIST, SRM.