Terbium standard solution, for ICP, in HNO₃, CRM, VHG
Lieferant: LGC Standards
PROOVHG-PTBN-100EA 51 EUR
PROOVHG-PTBN-100 PROOVHG-LTBN-100 PROOVHG-TTBN-100 PROOVHG-TTBN-500 PROOVHG-PTBN-500
Terbium standard solution, for ICP, in HNO₃, CRM, VHG
Standards Kationischer Standards Terbium-Standards
These solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Standards: Traceable to NIST, SRM.