Water standards for oils, Karl Fischer titration, CRM, VHG
Lieferant: LGC Standards
PROOVHG-KF-0.1P100EA 242 EUR
PROOVHG-KF-0.1P100 PROOVHGKF0.05P100 PROOVHGKF-0.5P-100 PROOVHG-KF-1.0P100
Water standards for oils, Karl Fischer titration, CRM, VHG
Standards Standards für die Wasserbestimmung (nach Karl Fischer)
This solution is intended for use as a certified reference material in the determination of water in motor oil by Karl Fischer titration.
Standards: Traceable to NIST and SRM.