Nitrate ion chromatography standard, VHG
Lieferant: LGC Standards
VHG
PROOVHG-INO3-100EA 56 EUR
PROOVHG-INO3-100 PROOVHG-I1PNO3-100 PROOVHG-INO3-500
Nitrate ion chromatography standard, VHG
Standards Anionische Standards Nitrat-Standards
This solution is intended for use as a certified reference material (CRM) and calibration standard, for ion chromatography (IC) and techniques using other modes of aqueous ion detection.
Standards: Traceable to NIST and SRM.