Bromide ion chromatography standards, bromide standard, VHG
Lieferant: LGC Standards
PROOVHG-IBR-500EA 110 EUR
PROOVHG-IBR-500 PROOVHG-IBR-100
Bromide ion chromatography standards, bromide standard, VHG
Standards Anionische Standards Bromid-Standards
These standard solutions are intended for use as a calibration or reference standard for ion chromatography (IC) or techniques using other modes of aqueous ion detection.
Standards: Traceable to NIST and CRM.