Reference materials, distillation standard, VHG
Lieferant: LGC Standards
VHG
PROOVHG-D86-4-500EA 0 EUR
PROOVHG-D86-4-500
Reference materials, distillation standard, VHG
Standards Mineralölindustrie-Standards
These solutions are intended for use as a distillation reference material for petroleum products at atmospheric pressure. This standard is a diesel oil whose distillation range is specified in ASTM D86 distillation Group 4.