BOD standard solutions, VHG
Lieferant: LGC Standards
PROOVHG-BODSEED-50EA 223 EUR
PROOVHG-BODSEED-50 PROOVHG-BOD200A
BOD standard solutions, VHG
Standards Umweltanalyse-Standards
The standard solution is intended for use as a glucose-glutamic acid standard check solution for biochemical oxygen demand (BOD) in APHA Method 5210 B.
Standards: Traceable to NIST.