Reference standards, surface water - trace metals
Lieferant: LGC Standards
PROOSPS-SW2EA 259 EUR
PROOSPS-SW2 PROOSPS-SW1
Reference standards, surface water - trace metals
Standards Umweltanalyse-Standards
This reference standard is intended for daily use in conjunction with measurement of elements in surface waters. The water solution contains 45 elements with concentrations traceable to
ultrapure metals or stoichiometrically well-defined substances.