Reference standards, PAH mixture, CERTAN®
Lieferant: Promochem
PROOSL30010EA 307 EUR
PROOSL30010 PROOSL30000
Reference standards, PAH mixture, CERTAN®
Standards Umweltanalyse-Standards
According to EU recommendation about the determination of carcinogenic PAHs in food (2005/108/EC) .