Turbidity standards, formazin
Lieferant: LGC Standards
PROORTCTURB10-500EA 131 EUR
PROORTCTURB10-500 PROORTCTURB100-500 PROORTCTURB05-500 PROORTCTURB1000100 PROORTCTURB1-500ML
Turbidity standards, formazin
Standards Trübungsstandards
Turbidity calibration standard formazin solutions, RTC ranging from 0,5 to 4000 NTU.