Organic reference standards, Multi-nutrient fertiliser - elements
Lieferant: LGC Standards
NIST
PROONIST-695EA 1420 EUR
PROONIST-695
Organic reference standards, Multi-nutrient fertiliser - elements
Standards Organische Standards
These standards are intended primarily for use in the evaluation of techniques employed in the analysis of multi-nutrient fertiliser materials and materials of a similar matrix.
One unit consists of approximately 70 g of jet-milled fertiliser.
Standards: Traceable to NIST and SRM.