Elemental analysis standards, portland cement (Ivory)
Lieferant: LGC Standards
PROONIST-1884BEA 1227 EUR
PROONIST-1884B
Elemental analysis standards, portland cement (Ivory)
Standards Umweltanalyse-Standards
These are intended primarily for the evaluation and calibration of methods for analysis of cements and materials of similar matrix.
A unit consists of five sealed vials, each containing approximately 4,5 g of portland cement ground to pass through a 75 µm (No. 200) sieve.
Standards: Traceable to NIST and SRM.