Tetrachlormethan-[13C] (99% ¹³C )

Lieferant: LGC Standards PROMOCHEM
Gefahr

PROOCILCLM-731-0.5EA 404 EUR
PROOCILCLM-731-0.5 PROOCIL-CLM-731-1 PROOCILCLM-731-0.1
Tetrachlormethan-[13C] (99% ¹³C )
Tetrachlormethan-[13C]
Formel: CCl₄
Lagertemperatur: Raumtemperatur
CAS-Nummer: 32488-50-9
UN: 1846
ADR: 6.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR