Organic reference standard, norethandrolone, Cerilliant

Lieferant: LGC Standards
PROOCERN-914EA 185 EUR
PROOCERN-914
Organic reference standard, norethandrolone, Cerilliant
Standards Organische Standards
  • Molecular formula: C₂₀H₃₀O₂
  • Molecular weight: 302,45 g/mol
  • CAS number: 52-78-8
  • Flash point: -20,0 °C
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR