Microplates, opaque OptiPlates™
Lieferant: PerkinElmer
PERK6005290EA 193 EUR
PERK6005290 PERK6005270 PERK6007290 PERK6005299
Microplates, opaque OptiPlates™
Mikrotiterplatten Zellkultur-Platten
These plates are used for general isotopic, fluorescence, LANCE™, [FP]², AlphaScreen™ or luminescence assays.
  • Made of polystyrene