Whole blood, ulcerative colitis (UC)
HCMSPB000F-2-UCEA 0 EUR
HCMSPB000F-2-UC
Whole blood, ulcerative colitis (UC)
Whole blood is used for researching the causes, diagnosis, supportive care and treatment of ulcerative colitis (UC). Collected from IRB-consented donors diagnosed with ulcerative colitis (UC).
No pre-screening is done on fresh products, but donors are tested on day of collection for Hepatitis B, Hepatitis C, and HIV 1/2, HTLV-I/II, HIV-1/HCV/HBV nucleic acid, WNV nucleic acid, Trypanasoma cruzi and Syphilis.
Storage: Use immediately upon receipt.
Bitte beachten Intended for research use only and not for direct therapeutic or diagnostic use in human or animals, or for further manufacturing of therapeutic products or devices.