Forceps, Axal™
Lieferant: Outils Rubis
Axal™
OUTI5-AXALEA 144 EUR
OUTI5-AXAL OUTI5A-AXAL OUTI7-AXAL OUTI0C-11-AXAL OUTI3C-AXAL OUTI2-AXAL OUTI7A-AXAL OUTI0-AXAL OUTISS-AXAL OUTI4-AXAL OUTI3-AXAL OUTI1-AXAL OUTI4AB-AXAL OUTIF-AXAL OUTI2A-AXAL
Forceps, Axal™
Pinzetten
Cobalt-chromium-molybdenum alloy. These forceps with very high Vicker hardness offer a range of physical, chemical and technological properties that make them appropriate for a wide variety of applications. Exceptional good tolerance on human tissues.
  • High yield strength alloy
  • 100% antimagnetic, rust proof and acid resistant
  • Highly resistant to corrosion, stress corrosion cracking and hydrogen embattlement
  • Temperature resistance of 500° C
  • Good fatigue and tensile strength guarantee durability
Perfect forceps for surgical appliances and laboratories.