AOCS Reference Mixtures, F.A.M.E. Mixes, certifed reference materials, Supelco®
Lieferant: Merck
TraceCERT®
O7381-1AMPEA 60.9 EUR
O7381-1AMP O7006-1AMP O7256-1AMP O7631-1AMP O7131-1AMP
AOCS Reference Mixtures, F.A.M.E. Mixes, certifed reference materials, Supelco®
Standards Mineralölindustrie-Standards
Quantitative animal and vegetable reference mixes which conform to the requirements of the American Oil Chemists′ Society (AOCS) method Ce 1-62.
Standards: These certified reference materials (CRM) are produced and certified in accordance with ISO 17034 and ISO/IEC 17025:2005.