Lysates, mammalian GenLysate™, human
Lieferant: G-Biosciences
GenLysate™
NLH-08EA 303 EUR
NLH-08 TLH-09 NLH-05 TLH-07 NLH-06 TLH-06 NLH-09 TLH-03 SLH-081 NLH-03 NLH-04 SLH-082 NLH-01 NLH-02 SLH-087 SLH-089 SLH-088 TLH-13 TLH-12 TLH-11 TLH-01 NLH-21 NLH-22 NLH-20 SLH-021 SLH-023 SLH-025 SLH-024 SLH-027 SLH-026 SLH-0210 SLH-029 SLH-028 NLH-18 NLH-19 TLH-19 NLH-16 NLH-17 TLH-18 TLH-17 TLH-16 TLH-15 TLH-14 SLH-090 SLH-0211 SLH-0212 NLH-11 SLH-0213 NLH-14 NLH-15 NLH-12 NLH-13 TLH-20
Lysates, mammalian GenLysate™, human
Zellen
GenLysate™ total protein lysates are ready to use for SDS-PAGE and Western blotting purposes. The total proteins are being extracted from whole tissue or cells following the method of Laemmli. The proteins are extracted in a denaturing buffer containing 62 mM Tris HCl and 2% SDS with ProteaseARREST™ protease inhibitor cocktail. This minimises proteolytic damage to the proteins.
GenLysate™ total protein lysates are offered as a 150 µg lyophilised protein per vial and, following reconstitution in DI water, are ready for use. GenLysate™ is also available in a wide variety of other species and tissues. Species include: Human (both normal tissue and tumour tissue lysates available), mouse, rat, rabbit, pig, plant-related, bacteria, insect, bird, frog, fish, marsupial, earthworm, and yeast.