RABS/isolator mittens, BioClean™ Sterile Nitrile
Lieferant: Ansell-Nitritex
BioClean™
112-5409EA 401 EUR
112-5409 NITRGGL30NITM9
RABS/isolator mittens, BioClean™ Sterile Nitrile
Handschuhe
These sterile RABS and isolator mittens are manufactured from nitrile with incredibly low levels of particles and excellent ESD properties.
  • Tested against ASTM D6978-05 for handling chemo drugs
  • 100% air leak tested
  • Suitable for autoclaving
  • Gamma irradiated (Sterility Assurance Level (SAL) of 10⁻⁶)
  • Can be sanitised by VHP or IPA
  • Ultra-clean surface ensures product protection
  • Specially designed to minimise hand fatigue
Designed for use in product contact areas, the GGL mittens are fully validated for sterility with an SAL (Sterility Assurance Level) of 10⁻⁶.
Suitable for ISO Class 4 and EU GMP Grade A cleanrooms.
Standards: EN 388:2016 (4101X), EN ISO 374-1 Type A (AJKNOT), EN ISO 374-5 VIRUS, EN 421; Cat. III
Food contact approved
Tested against permeation standard ASTM D6978-05 for cytotoxic drugs.
Verpackung: Triple-bagged: One piece per sealed inner PE bag; one inner bag per sealed second inner PE bag; one second inner bag per sealed outer PE bag; 20 outer bags per lined inner white Correx polyethylene box (20 pieces).
Bitte beachten No glove provides complete protection against all chemicals. Users must test the gloves against the particular chemicals and environment in which they will be used.