Block heaters, analogue, Thermubloc, BH-1T series
Lieferant: Nickel-Electro
NICKBH-1TEA 961 EUR
NICKBH-1T
Block heaters, analogue, Thermubloc, BH-1T series
Clifton® block heaters provide a safe, dry and consistent method of heating accessory aluminium blocks with excellent thermal conductivity for heating tubes. Features a thermal over temperature protection, chemical resistant control panel, corrosion resistent stainless steel case and anti-bacterial technology.
  • The paint finish actively inhibits bacterial growth improving hygiene requirements as demanded by many clinical and laboratory environments
  • This anti-bacterial agent is integral within the paint finish
Lieferumfang: Supplied with block extraction tool.
Bitte beachten * Capacity is 1 block if 10; 12; 13; 16; 19; 25 mm or 50 ml tubes and 2 blocks if 0,2; 0,5; 1,5 or 2 ml tubes.