Bacterial freezing media

Lieferant: VWR Life Science

VWR®
N400-25PKEA 364 EUR
N400-25PK
Bacterial freezing media
Zelleinfriermedien
Bacterial freezing media is a sterile, ready to use cryopreservation media for long term storage of bacterial cultures. Ideal for the preservation of transformed bacteria as well as non fastidious bacteria.

  • Each vial contains 0,5 ml LB broth (Miller formulation) with 30% glycerol
  • Vials are marked at 0,5 ml increments and provide an area for recording strain information
  • Viability of frozen cultures after thawing is 85%

Add 1 ml of saturated culture to the vial, vortex, and store at –70 °C

Lieferumfang: Supplied in a convenient storage box containing 25 single use, 2 ml capacity vials.
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR