Rapid aerobic count plates, Petrifilm™
Lieferant: 3M
MMMP6478EA 142 EUR
MMMP6478 MMMP6479
Rapid aerobic count plates, Petrifilm™
Medien für die Mikrobiologie Fertigmedien für die Mikrobiologie Fertigmedien auf Petrifilm™
3M™ Petrifilm™ Rapid aerobic count plates feature technology that facilitates colony enumeration in just 24 hours for most food matrices/environmental samples and resists distortion caused by spreader colonies.
  • Provides results as soon as 24 hours (excluding dairy powders) of incubation time
  • Plates are sample-ready, eliminating the time-consuming, cumbersome step of preparing media/agar dishes
  • Can be used with the 3M™ Petrifilm™ Plate reader for consistent, fast, automated reading and recording of results
  • Dual-sensing indicator technology that facilitate colony enumeration
  • Food processors and contract labs worldwide trust 3M™ Petrifilm Plates to provide accurate, efficient, consistent results
Aerobic count bacteria are present in a variety of food matrices and serve as indicators for food spoilage. Getting accurate aerobic bacteria counts in your raw materials, finished product and production environment are critical to making time sensitive decisions that impact process control, cleaning decisions and, ultimately, product quality and safety. 3M™ Petrifilm™ Rapid aerobic count plates are a sample ready, culture medium system, which contain nutrients, a coldwater soluble gelling agent, and a dual sensing indicator technology that facilitate colony enumeration in 24 hours for most food matrices and environmental samples. Proven as reliable as the SMA plate method, 3M™ Petrifilm Rapid aerobic count plates help agar users avoid costly retesting and delayed results due to interpretation issues caused by spreader colonies. The 3M™ Petrifilm Rapid aerobic count plates can also be read using the 3M™ Petrifilm Plate reader which provides consistent, fast, automated reading and recording of results of 3M™ Petrifilm plates (aerobic count, Rapid aerobic count, Coliform count, E. coli/Coliform count, Enterobacteriaceae count and select E. coli count ) in 4 seconds.
The 3M™ Petrifilm plate advantage in today’s environment of heightened focus on food safety and increasingly stringent quality requirements, labs are under pressure to provide tests that are fast, reliable and consistently accurate. 3M™ Petrifilm plates have demonstrated consistent performance that has been cited globally with more than 200 certificates, recognitions, validations, and peer review publications. For more than 30 years, food safety professionals around the world have put their trust in 3M™ Petrifilm plates. That’s because 3M™ Petrifilm plates have streamlined, standardised and simplified the process of microbial quantitative indicator testing, improving productivity and helping ensure the highest levels of product quality.