Partikelfilternde Halbmasken
Lieferant: 3M Medical
MMMEM1872VEA 101 EUR
MMMEM1872V MMMEM1862 MMMEM1873V MMMEM1861
Partikelfilternde Halbmasken
Atemschutzmasken Einweg-Atemschutzmasken Atemschutzmasken gegen Partikel
The 1800 Series offers the wearer comfortable and reliable protection. The foldable 3-panel design fits a wide range of face shapes and is compatible with eyewear.
  • Nose foam made from sweat absorbent material to avoid slippage on face
  • 3M™ Advanced Electret Filter Material ensures effective filtration and low breathing resistance for comfortable wear
  • 3M™ Cool Flow™ Valve removes heat and moisture build up and minimises the risk of misting eyewear (model 1872 and 1873)
  • Colour coded to indicate performance level: Yellow FFP1, blue FFP2, red FFP3
  • Hygienic packaging protects the respirator from contamination before use and during storage
  • Latex free
Standards: EN 149:2001