P3 R Hartschalen-Partikelfilter, Secure Click™
Lieferant: 3M
Secure Click™
111-2570EA 456 EUR
111-2570
P3 R Hartschalen-Partikelfilter, Secure Click™
Atemschutzmasken Mehrweg-Atemschutzmasken Halbmasken
This series introduces an alternative to the 3M round bayonet connector with the new Secure Click™ connection, resulting in a quick, simple filter connection. It works just like a seat belt: Simply align the connections, push until audible click, and feel confident that filters are installed properly.