1,2,3,4,5-Pentabrom-6-chlorbenzol

Lieferant: Thermo Scientific

MAYBBTB15046.250EA 0 EUR
MAYBBTB15046.250 MAYBBTB15046.1
1,2,3,4,5-Pentabrom-6-chlorbenzol
1,2,3,4,5-Pentabrom-6-chlorbenzol
Formel: C₆Br₅Cl
CAS-Nummer: 13075-05-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR