TLC education advanced kit, micro-set F3
Lieferant: Macherey-Nagel
MANA814402EA 35.1 EUR
MANA814402 MANA814403 MANA814400 MANA814401 MANA814406 MANA814407 MANA814404 MANA814405
TLC education advanced kit, micro-set F3
Lehrmittel Chemie-Lehrmittel Chromatographie-Lehrmittel
Advanced kit for science education.
  • All chemicals included for the separation of:
  • Analgetics (pain relievers)
  • Drugs as shown for cinchona bark
Together with the materials from TLC micro-Set M Aspirin® and Thomapyrin® tablets as well as the drug cinchona bark can be separated and visualised by spray reagents (e.g., reagent according to Dragendorff-Munier).
Bitte beachten * These products contain harmful substances which must be specially labelled as hazardous.