GC injection port septa
Lieferant: Macherey-Nagel
MANA702660EA 68.2 EUR
MANA702660 MANA702661 MANA702619 MANA702659 MANA702613 MANA702657 MANA702658 MANA702611 MANA702656 MANA702612 MANA702610 MANA702632 MANA702609 MANA702628 MANA702606 MANA702648 MANA702604 MANA702626 MANA702605 MANA702649 MANA702627 MANA702646 MANA702602 MANA702647 MANA702625 MANA702622 MANA702623 MANA702620 MANA702621 MANA702662 MANA702663
GC injection port septa
Septen
GC septa with premium HT and standard quality.
Premium HT quality:
- Blister pack for cleanliness and easily handling
- CenterGuide™ for a centred needle insertion
- High inert and durable materials
- High temperature stability (300/400 °C)
Standard quality:
- Inert silicone materials
- Temperature stability from 200 to 320 °C
* Plug