Screw neck vials, N 8
Lieferant: Macherey-Nagel
MANA702251EA 44.2 EUR
MANA702251 MANA70213.2 218-1105 MANA702248 MANA702249 MANA702238 MANA702246 MANA702004 MANA702247 MANA702860 MANA702893
Screw neck vials, N 8
Vials
These screw neck vials are characterised by a small opening and 8-425 thread.
  • Made of clear and amber glass
  • Available with flat or conical bottom
  • Different inserts available
Often used on instruments of Dionex (Thermo Scientific), Knauer, Thermo Scientific, Varian (Agilent), VWR® (Merck / Hitachi) and others.
Vials with closed top screw closures are suitable for sample storage.
The suitable screw closures are ready assembled.
- Septa made from economical Red Rubber/FEP, high quality silicone/PTFE or from virgin PTFE
- Silicone/PTFE septa are also slitted
Also available as convenient vial kits with 100 vials and 100 caps.