Sedimentiergefäße, Imhoff
Lieferant: Lenz
LENZ05670005EA 213.33 EUR
LENZ05670005 LENZ05670003 LENZ05670001
Sedimentiergefäße, Imhoff
Trichter Sedimentiergefäße
Made of borosilicate glass 3.3.
  • White graduations
  • With or without stopcock
  • Excellent chemical and temperature resistance