Scheidetrichter, Squibb
Lieferant: Lenz
LENZ04018037EA 136 EUR
LENZ04018037 LENZ04018049 LENZ04018137 LENZ04018058 LENZ04018149
Scheidetrichter, Squibb
Trichter Scheidetrichter
Separating funnels, Squibb, stopcock with glass or PTFE key, made of borosilicate glass amberized.
  • Without graduation
  • Very good temperature and chemical resistance
  • Amber coloured for ligth sensitve liquids