Drums, wide mouth, UN approved
Lieferant: Lamaplast
LAMA1363EA 184 EUR
LAMA1363 LAMA1352 LAMA1364 LAMA1353 LAMA1361 LAMA1351
Drums, wide mouth, UN approved
Fässer, Fässer, Trommeln
Drums, made of HDPE, blue, UN approved, with lid.
  • Stackable
  • Tamper evident
  • Self-sealing lid with zinc plate steel closure
  • For food use
For food, cosmetic and chemical fields. Manufactured with the inner neutral layer that avoids the contact between the content and the coloring pigment.
Dimensions allow overlap on pallet 800×1200 and 1000×1200.