Microplate fluorometer and luminometer, Fluoroskan™ FL
Lieferant: Thermo Fisher Scientific
LABL5200220EA
22580
EUR
LABL5200220
LABL5200221
LABL5200222
Microplate fluorometer and luminometer, Fluoroskan™ FL
The Thermo Scientific™ Fluoroskan™ FL is a compact and robust microplate fluorometer and luminometer, offering simplicity and reliability for increased efficiency of workflows. Featuring excellent optical performance, the Fluoroskan™ FL Microplate Fluorometer and Luminometer is ideal for life science fluorometric and luminometric assays. The Fluoroskan™ FL fluorometer and luminometer offers plate format versatility, fast plate reading speeds, and up to three dispensers.
- High sensitivity for both top and bottom reading
- Fast reading speed essential for kinetic applications
- Onboard dispensers for precise initiation of kinetic reactions
- Powerful and easy-to-use Thermo Scientific™ SkanIt™ software
The use of fibreless direct illumination optics for both top and bottom reading enables high sensitivity, a wide dynamic range, and accurate and precise results for 6- to 384-well plates.
The Fluoroskan™ FL Microplate Fluorometer and Luminometer is capable of reading a 96-well plate in just 18 seconds, essential for kinetic applications. Fluorometric GFP measurement and luminometric luciferase measurement from the same well occurs quickly due to the versatility of the assay programming.
The Fluoroskan™ FL Microplate Fluorometer and Luminometer can be equipped with up to three reagent dispensers* for use during fast kinetic assays, such as Ca2+ flux assays. The onboard dispensers support simultaneous dispensing and reading, enabling the monitoring of fast kinetic measurements from the very start of the reaction. Very low dead volume and backflush capability support conservation of reagents.
With a visual workflow setup, SkanIt™ software is quick and easy to learn. It offers effortless data analysis (with a comprehensive selection of built-in calculations, including blank subtraction, standard curve creation, classification, quality control, kinetic analysis) and exporting capabilities. Each user can install SkanIt™ software on his/her own computer to set up assays and analyse data.
The Fluoroskan™ FL Microplate Fluorometer and Luminometer is capable of reading a 96-well plate in just 18 seconds, essential for kinetic applications. Fluorometric GFP measurement and luminometric luciferase measurement from the same well occurs quickly due to the versatility of the assay programming.
The Fluoroskan™ FL Microplate Fluorometer and Luminometer can be equipped with up to three reagent dispensers* for use during fast kinetic assays, such as Ca2+ flux assays. The onboard dispensers support simultaneous dispensing and reading, enabling the monitoring of fast kinetic measurements from the very start of the reaction. Very low dead volume and backflush capability support conservation of reagents.
With a visual workflow setup, SkanIt™ software is quick and easy to learn. It offers effortless data analysis (with a comprehensive selection of built-in calculations, including blank subtraction, standard curve creation, classification, quality control, kinetic analysis) and exporting capabilities. Each user can install SkanIt™ software on his/her own computer to set up assays and analyse data.
Applications include: Ca2+ flux assays , cell proliferation, cytotoxicity, cell adhesion, nucleic acid quantitation, reporter gene assays, hybridization assays, ion channel, FRET assays, BRET assays, immunoassays, enzyme activity, bacterial quantitation, phagocytosis, oligonucleotide assays.
Bestellinformation: Includes SkanIt™ PC Software and the following filter pairs (excitation/emission): 355 nm/460 nm, 485 nm/538 nm. An extensive selection of additional filters is available upon request.
Bitte beachten *Upgrade to third dispenser must be performed by service technician
Neuer Artikel bei VWR!
Limited Time Sale
Select items below are currently on sale
Artikel wurde zum Warenkorb hinzugefügt