4-Amino-1-Boc-piperidin-4-carbonsäure 98%
Lieferant: Thermo Scientific
L19794.03EA
176
EUR
L19794.03
L19794.MD
4-Amino-1-Boc-piperidin-4-carbonsäure 98%
4-Amino-1-Boc-piperidin-4-carbonsäure
Formel:
C₁₁H₂₀N₂O₄ Molecular Weight: 244,29 g/mol Schmelzpunkt: ∼ 290 °C (Zersetzung) |
MDL:
MFCD01318728 CAS-Nummer: 183673-71-4 EINECS: 000-000-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...