4-(Trifluoracetyl)benzoesäure ≥97%

Lieferant: Thermo Scientific
Achtung

L10242.03EA 91.1 EUR
L10242.03 L10242.MD
4-(Trifluoracetyl)benzoesäure ≥97%
4-(Trifluoracetyl)benzoesäure
Formel: C₉H₅F₃O₃
Molecular Weight: 218,13 g/mol
Schmelzpunkt: 177…179 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00052340
CAS-Nummer: 58808-59-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR