Pulvertrichter
Lieferant: Bohemia Cristal (Kavalierglass)
KAVA632413102100EA 67 EUR
KAVA632413102100 KAVA632413102080 KAVA632413102060
Pulvertrichter
Trichter Pulvertrichter
Borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
These funnels with short stem and bevelled edges are suitable for transferring powdery materials.
Bestellinformation: Ø = internal stem diameter.