Bunsen burner
Lieferant: Juchheim
JUCHVL01492PEA 351 EUR
JUCHVL01492P JUCHVL01492E
Bunsen burner
Heizgeräte Gasbrenner
Bunsen burner with foot switch and magnetic valve.
Height: 145 mm
Weight: 1055 g
Height: 145 mm
Weight: 1055 g