Teclu safety burner
Lieferant: Juchheim
JUCHVL01374DPEA 148.4 EUR
JUCHVL01374DP JUCHVL01374P JUCHVL01374DE JUCHVL01374E
Teclu safety burner
Heizgeräte Gasbrenner
Teclu safety burner with needle valve and thermofeeler with safety valve on heavy, round base.
Height: 200 mm
Weight: 685 g