Magnetic stirrer heads with conical joint
Lieferant: Juchheim
JUCH10264.00EA 1565.4 EUR
JUCH10264.00 JUCH10262.00
Magnetic stirrer heads with conical joint
Rührkupplungen
Conical joint stirrer heads made of stainless steel bottom, special steel shaft, chromium plated brass upper body for vacuum and slight positive pressure.