These safety exhaust filters can be placed on the waste bottle of an HPLC system, to protect workers from solvent vapors in the ambient air.
- Prevents pollution of ambient air with volatile compounds
- Break-through detector alerts the users about the saturation of the filter absorbent
- Universal 1/4-28 connection
The break-through detector recognises trace amounts of acetone, acetonitrile, ethanol, ethyl acetate, methanol, methylene chloride and THF. When the absorbent is saturated, break-through is indicated by a colour change in the detector from orange/pink to dark purple. A small charcoal filter absorbs the vapours after they pass through the detector.