Safety exhaust filters
Lieferant: VICI Jour
JOURJR-S-20005EA 273 EUR
JOURJR-S-20005 JOURJR-S-20002
Safety exhaust filters
Filter Belüftungsfilter
These safety exhaust filters can be placed on the waste bottle of an HPLC system, to protect workers from solvent vapors in the ambient air.
  • Prevents pollution of ambient air with volatile compounds
  • Break-through detector alerts the users about the saturation of the filter absorbent
  • Universal 1/4-28 connection
The break-through detector recognises trace amounts of acetone, acetonitrile, ethanol, ethyl acetate, methanol, methylene chloride and THF. When the absorbent is saturated, break-through is indicated by a colour change in the detector from orange/pink to dark purple. A small charcoal filter absorbs the vapours after they pass through the detector.