Yeast nitrogen base (YNB), VWR®
Lieferant: VWR Collection
J386-500GEA 1050 EUR
J386-500G J630-100G J630-500G J386-100G
Yeast nitrogen base (YNB), VWR®
Medien für die Mikrobiologie Basismaterialien für die Mikrobiologie
Yeast Nitrogen Base (YNB) is a base medium for preparation of minimal and synthetic defined yeast media.
  • Animal origin
  • Suitable for studying strains of yeast requiring certain vitamins
Bestellinformation: Base materials are available with or without ammonium sulfate for the classification of yeast.