Taq CORE kits
Lieferant: MP Biomedicals
ICNA11EPTQK10LEA 2536 EUR
ICNA11EPTQK10L ICNA11EPTQK255C ICNA11EPTQKD259 ICNA11EPTQK25L ICNA11EPTQK105C ICNA11EPTQK300 ICNA11EPTQK101 ICNA11EPTQK103 ICNA11EPTQK251 ICNA11EPTQKD251 ICNA11EPTQKL255 ICNA11EPTQKD253 ICNA11EPTQK253 ICNA11EPTQK109 ICNA11EPTQK259 ICNA11EPTQS901K
Taq CORE kits
Nukleinsäure-Reagenzien Enzyme und Kits für Endpunkt-PCR
The Taq CORE kits are designed for the amplification of specific regions of DNA using PCR. The PCR CORE Kits - Direct Loading provide Taq DNA Polymerase, PCR Direct Loading buffers and dNTPs necessary for amplification and subsequent loading of samples on an agarose gel.
  • Contain dNTPs at 10 mM
  • Highly reproducible results
  • Contains ultra-pure enzyme