Taq DNA polymerase
Lieferant: MP Biomedicals
ICNA11EPTQA925EA 1281 EUR
ICNA11EPTQA925 ICNA11EPTQX925 ICNA11EPTQX025 ICNA11EPTQL100 ICNA11EPTQC060 ICNA11EPTQA100 ICNA11EPTQX100 ICNA11EPTQD325 ICNA11EPTQA025 ICNA11EPTQC200 ICNA11EPTQD025 ICNA11EPTQA325 ICNA11EPTQA105 ICNA11EPTQD925
Taq DNA polymerase
Nukleinsäure-Reagenzien Enzyme und Kits für Endpunkt-PCR
Taq DNA Polymerase is a specialised thermostable enzyme isolated from the thermophilic bacterium Thermus aquaticus. The recombinant form of this enzyme is expressed in E. coli. It is highly processive, recombinant polymerase which catalyses the addition of mononucleotide units to the 3’-OH end of a primer chain. It remains functional even after prolonged incubation at 95 °C. This enzyme possesses 5’-3’ exonuclease activiy.
Taq DNA Polymerase is used for the DNA polymerase chain reaction (PCR) in order to amplify DNA sequences. Other applications include RT-PCR, DNA sequencing and DNA labeling.