PCR master mix, Taq-&LOAD™
Lieferant: MP Biomedicals
Taq-&GO™
ICNA11EPTAL110EA 882 EUR
ICNA11EPTAL110 ICNA11EPTAL100
PCR master mix, Taq-&LOAD™
Nukleinsäure-Reagenzien Enzyme und Kits für Endpunkt-PCR
Taq-&LOAD™ PCR Master Mix is an optimised 5X reaction mix for PCR containing recombinant Taq DNA polymerase, dNTPs, MgCl₂, and incubation and loading buffers. Also included are a compound increasing the density of the sample and a red/purple dye.
  • Significant savings in preparation time
  • Reduced pipetting
  • Reduced risk of errors and contamination
  • Stable at 4 °C
This mastermix allows direct loading onto the agarose gel after PCR without the need for additional loading dye. The included red purple dye migrates at 400 bp on a 1,2% agarose gel.